(1S)-(-)-Camphanic chloride 95%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
(1S)-4,7,7-Trimethyl-3-oxo-2-oxabicyclo[2.2.1]heptane-1-carbonyl chloride, (-)-Camphanic acid chloride
L14148.03
L14148.06
ALFAL14148.03EA
69.5
SGD
ALFAL14148.03
ALFAL14148.06
(1S)-(-)-Camphanic chloride 95%
(1S)-(-)-Camphanic chloride
Purity: 95%
CAS No.: 39637-74-6
Molecular Formula: C10H13ClO3
Formula Weight: 216.67
Storage Temperature: -30°C to -10°C
Melting Point: 64-68°
Beilstein Reference: 3590860
Optical Rotation: -18° (c=2 in carbon tetrachloride)
UN No.: 3261
MDL No.: MFCD00135626
Formula:
C₁₀H₁₃ClO₃ MW: 216,67 g/mol Melting Pt: 64…68 °C |
MDL Number:
MFCD00135626 CAS Number: 39637-74-6 EINECS: 254-552-4 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...