Propargyltriphenylphosphonium bromide ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: 2-Propinyl-triphenylphosphonium bromide, Triphenyl(prop-2-yn-1-yl)phosphonium bromide

A12753.06 A12753.14 A12753.22
ALFAA12753.06EA 0 SGD
ALFAA12753.06 ALFAA12753.14 ALFAA12753.22
Propargyltriphenylphosphonium bromide ≥97%
Propargyltriphenylphosphonium bromide

Purity: 97% CAS No.: 2091-46-5 Molecular Formula: C21H18BrP Formula Weight: 381.26 Odour: Odourless Melting Point: 177-182° Beilstein Reference: 3579775 MDL No.: MFCD00011811

Formula: C₂₁H₁₈BrP
MW: 381,26 g/mol
Melting Pt: 177…182 °C
Storage Temperature: Ambient
MDL Number: MFCD00011811
CAS Number: 2091-46-5
EINECS: 218-242-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR