1,2-Bis(dicyclohexylphosphino)ethane ≥98%
Supplier: Thermo Scientific
|
Danger
|
Packaged under argon.
39269.02
39269.04
ALFA39269.02EA
0
SGD
ALFA39269.02
ALFA39269.04
1,2-Bis(dicyclohexylphosphino)ethane ≥98%
1,2-Bis(dicyclohexylphosphino)ethane
Purity: 98%
CAS No.: 23743-26-2
Molecular Formula: (C6H11)2PCH2CH2P(C6H11)2
Formula Weight: 422.61
Physical Form: Crystalline, Packaged under argon
Sensitivity: Air Sensitive
UN No.: 1325
MDL No.: MFCD00015521
Formula:
C₂₆H₄₈P₂ MW: 422,607 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00015521 CAS Number: 23743-26-2 UN: 1325 ADR: 4.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...