3-Fluoro-2-nitropyridine 96%
Supplier: Thermo Scientific
|
Warning
|
H33813.03
H33813.06
ALFAH33813.03EA
328
SGD
ALFAH33813.03
ALFAH33813.06
3-Fluoro-2-nitropyridine 96%
3-Fluoro-2-nitropyridine
Formula:
C₅H₃FN₂O₂ Melting Pt: 30…35 °C Storage Temperature: Ambient |
MDL Number:
MFCD04114127 CAS Number: 54231-35-5 EINECS: 207-260-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...