Tin arsenide ≥99.999% (metals basis)

Supplier: Thermo Scientific
Danger

36363.06 36363.14
ALFA36363.06EA 0 SGD
ALFA36363.06 ALFA36363.14
Tin arsenide ≥99.999% (metals basis)
Tin arsenide

Purity: 99.999% CAS No.: 39332-13-3 Molecular Formula: SnAs Formula Weight: 193.61 Physical Form: 6mm & down pieces UN No.: 1557 MDL No.: MFCD00148646

Formula: As₄Sn₃
MW: 193,61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00148646
CAS Number: 39332-13-3
UN: 1557
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR