4-Iodonicotinonitrile 95%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
3-Cyano-4-iodopyridine
H27603.MD
H27603.03
ALFAH27603.MDEA
0
SGD
ALFAH27603.MD
ALFAH27603.03
4-Iodonicotinonitrile 95%
4-Iodonicotinonitrile
Purity: 95%
CAS No.: 490039-72-0
Molecular Formula: C6H3IN2
Formula Weight: 230.01
Storage Temperature: -30°C to -10°C
Melting Point: ca 124° dec.
UN No.: 3439
MDL No.: MFCD09037453
Formula:
C₆H₃IN₂ MW: 230,01 g/mol Melting Pt: ∼ 124 °C (dec.) |
MDL Number:
MFCD09037453 CAS Number: 490039-72-0 UN: 3439 ADR: 6.1,III |
Learn more
![](/m-sg.vwr.com/en_SG/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...