1-Bromo-3,3,4,4,4-pentafluoro-2-butanone ≥97%

Supplier: Thermo Scientific
Danger

L12199.03 L12199.06
ALFAL12199.03EA 0 SGD
ALFAL12199.03 ALFAL12199.06
1-Bromo-3,3,4,4,4-pentafluoro-2-butanone ≥97%
1-Bromo-3,3,4,4,4-pentafluoro-2-butanone

Purity: 97% CAS No.: 92737-01-4 Molecular Formula: C4H2BrF5O Formula Weight: 240.96 Boiling Point: 93-94° UN No.: 3265 MDL No.: MFCD00041447

Formula: C₄H₂BrF₅O
MW: 240,96 g/mol
Boiling Pt: 93…94 °C
Storage Temperature: Ambient
MDL Number: MFCD00041447
CAS Number: 92737-01-4
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR