4-(Methylamino)pyridine 99%

Supplier: Thermo Scientific
Warning

L19626.03 L19626.06 L19626.14
ALFAL19626.03EA 104 SGD
ALFAL19626.03 ALFAL19626.06 ALFAL19626.14
4-(Methylamino)pyridine 99%
4-(Methylamino)pyridine

Purity: 99% CAS No.: 1121-58-0 Molecular Formula: C6H8N2 Formula Weight: 108.14 Melting Point: 124-128° Beilstein Reference: 108213

Formula: C₆H₈N₂
MW: 108,14 g/mol
Melting Pt: 124…128 °C
Storage Temperature: Ambient
MDL Number: MFCD00152625
CAS Number: 1121-58-0
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR