5-Bromo-1-isoindolinone 96%

Supplier: Thermo Scientific

Synonyms: 5-Bromo-2,3-dihydro-1H-isoindol-1-one, 5-Bromoisoindolin-1-one

H52358.MD H52358.03
ALFAH52358.MDEA 264 SGD
ALFAH52358.MD ALFAH52358.03
5-Bromo-1-isoindolinone 96%
5-Bromo-1-isoindolinone

Purity: 96% CAS No.: 552330-86-6 Molecular Formula: C8H6BrNO Formula Weight: 212.04 MDL No.: MFCD09701292

Formula: C₈H₆BrNO
MW: 212,04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09701292
CAS Number: 552330-86-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR