3-(Trifluoromethyl)benzhydrol ≥97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Phenyl(3-(trifluoromethyl)phenyl)methanol
L14414.03
L14414.06
ALFAL14414.03EA
0
SGD
ALFAL14414.03
ALFAL14414.06
3-(Trifluoromethyl)benzhydrol ≥97%
3-(Trifluoromethyl)benzhydrol
Purity: 97%
CAS No.: 728-80-3
Molecular Formula: C14H11F3O
Formula Weight: 252.23
Melting Point: 62-64°
Boiling Point: 125-127°/0.5mm
MDL No.: MFCD00014397
Formula:
C₁₄H₁₁F₃O MW: 252,23 g/mol Boiling Pt: 125…127 °C (0,5 mmHg) Melting Pt: 62…64 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014397 CAS Number: 728-80-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...