2-Phosphonobutyric acid ≥97%

Supplier: Thermo Scientific
Danger

L12036.09 L12036.04
ALFAL12036.09EA 512 SGD
ALFAL12036.09 ALFAL12036.04
2-Phosphonobutyric acid ≥97%
2-Phosphonobutyric acid

Purity: 97% CAS No.: 4378-40-9 Molecular Formula: C4H9O5P Formula Weight: 168.09 Odour: Odourless Melting Point: 123-127° UN No.: 3261 MDL No.: MFCD00041416

Formula: C₄H₉O₅P
MW: 168,09 g/mol
Melting Pt: 123…127 °C
Storage Temperature: Ambient
MDL Number: MFCD00041416
CAS Number: 4378-40-9
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR