Tetramethylenebis(triphenylphosphonium bromide) ≥98%
Supplier: Thermo Scientific
|
Warning
|
A14214.09
A14214.18
ALFAA14214.09EA
136
SGD
ALFAA14214.09
ALFAA14214.18
Tetramethylenebis(triphenylphosphonium bromide) ≥98%
Tetramethylenebis(triphenylphosphonium bromide)
Purity: 98+%
CAS No.: 15546-42-6
Molecular Formula: Br(C6H5)3P(CH2)4P(C6H5)3Br
Formula Weight: 740.51
Melting Point: 292-293°
Beilstein Reference: 3872522
Formula:
C₄₀H₃₈Br₂P₂ MW: 740,51 g/mol Melting Pt: 292…293 °C Storage Temperature: Ambient |
MDL Number:
MFCD00031714 CAS Number: 15546-42-6 EINECS: 239-598-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...