1-Methylimidazole-5-carboxylic acid methyl ester ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Methyl 1-methylimidazole-5-carboxylate, Methyl 1-methyl-1H-imidazole-5-carboxylate
44688.01
44688.03
ALFA44688.01EA
0
SGD
ALFA44688.01
ALFA44688.03
1-Methylimidazole-5-carboxylic acid methyl ester ≥98%
1-Methylimidazole-5-carboxylic acid methyl ester
Purity: 98%
CAS No.: 17289-20-2
Molecular Formula: C6H8N2O2
Formula Weight: 140.14
Physical Form: Powder
Appearance: Pale yellow
Melting Point: 56-58°
MDL No.: MFCD01567300
Formula:
C₆H₈N₂O₂ MW: 140,14 g/mol Melting Pt: 56…58 °C Storage Temperature: Ambient |
MDL Number:
MFCD01567300 CAS Number: 17289-20-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...