1-Methylimidazole-4-carboxylic acid 95%

Supplier: Thermo Scientific
Warning

Synonyms: 1-Methyl-1H-imidazole-4-carboxylic acid

H51866.MD H51866.03
ALFAH51866.MDEA 93.1 SGD
ALFAH51866.MD ALFAH51866.03
1-Methylimidazole-4-carboxylic acid 95%
1-Methylimidazole-4-carboxylic acid

Purity: 95% CAS No.: 41716-18-1 Molecular Formula: C5H6N2O2 Formula Weight: 126.11 Melting Point: 241-243° MDL No.: MFCD02179560

Formula: C₅H₆N₂O₂
MW: 126,11 g/mol
Melting Pt: 241…243 °C
Storage Temperature: Ambient
MDL Number: MFCD02179560
CAS Number: 41716-18-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR