p-Xylylenebis(triphenylphosphonium bromide) 96%

Supplier: Thermo Scientific
Warning

A18241.14 A18241.22
ALFAA18241.14EA 0 SGD
ALFAA18241.14 ALFAA18241.22
p-Xylylenebis(triphenylphosphonium bromide) 96%
p-Xylylenebis(triphenylphosphonium bromide)

Purity: 96% CAS No.: 40817-03-6 Molecular Formula: C44H38Br2P2 Formula Weight: 788.55 Melting Point: 300° Beilstein Reference: 3584592

Formula: C₄₄H₃₈Br₂P₂
MW: 788,55 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00011914
CAS Number: 40817-03-6
EINECS: 255-092-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR