8-Quinolinesulphonyl chloride ≥97%

Supplier: Thermo Scientific
Danger

Synonyms: Quinoline-8-sulphonyl chloride

L05676.09 L05676.18
ALFAL05676.09EA 0 SGD
ALFAL05676.09 ALFAL05676.18
8-Quinolinesulphonyl chloride ≥97%
8-Quinolinesulphonyl chloride

Purity: 97% CAS No.: 18704-37-5 Molecular Formula: C9H6ClNO2S Formula Weight: 227.67 Melting Point: 127-130° Boiling Point: 306° UN No.: 3261 MDL No.: MFCD00006808

Formula: C₉H₆ClNO₂S
MW: 227,67 g/mol
Boiling Pt: 306 °C
Melting Pt: 127…130 °C
Storage Temperature: Ambient
MDL Number: MFCD00006808
CAS Number: 18704-37-5
EINECS: 242-515-5
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR