5-Bromo-3-iodo-2-methoxypyridine ≥97%

Supplier: Thermo Scientific
Warning

H50061.03 H50061.06
ALFAH50061.03EA 325 SGD
ALFAH50061.03 ALFAH50061.06
5-Bromo-3-iodo-2-methoxypyridine ≥97%
5-Bromo-3-iodo-2-methoxypyridine

Purity: 97% CAS No.: 578007-66-6 Molecular Formula: C6H5BrINO Formula Weight: 313.92 MDL No.: MFCD05663522

Formula: C₆H₅BrINO
MW: 313,92 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05663522
CAS Number: 578007-66-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR