(2-Chloro-4-pyridinyl)methanol ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: (2-Chloropyridin-4-yl)methanol, 2-Chloro-4-(hydroxymethyl)pyridine, 2-Chloropyridine-4-methanol

H50099.03 H50099.06
ALFAH50099.03EA 0 SGD
ALFAH50099.03 ALFAH50099.06
(2-Chloro-4-pyridinyl)methanol ≥97%
(2-Chloro-4-pyridinyl)methanol

Purity: 97% CAS No.: 100704-10-7 Molecular Formula: C6H6ClNO Formula Weight: 143.57 Melting Point: 64-66° MDL No.: MFCD06858767

Formula: C₆H₆ClNO
MW: 143,57 g/mol
Melting Pt: 64…66 °C
Storage Temperature: Ambient
MDL Number: MFCD06858767
CAS Number: 100704-10-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR