Methyl-3-methylbenzo[b]thiophene-2-carboxylate ≥97%

Supplier: Thermo Scientific

Synonyms: Methyl 3-methylbenzo[b]thiophene-2-carboxylate

B24069.03 B24069.06 B24069.14
ALFAB24069.03EA 0 SGD
ALFAB24069.03 ALFAB24069.06 ALFAB24069.14
Methyl-3-methylbenzo[b]thiophene-2-carboxylate ≥97%
Methyl 3-methylbenzo[b]thiophene-2-carboxylate

Purity: 97% CAS No.: 3133-81-1 Molecular Formula: C11H10O2S Formula Weight: 206.26 Melting Point: 102-105° MDL No.: MFCD02683110

Formula: C₁₁H₁₀O₂S
MW: 206,26 g/mol
Melting Pt: 102…105 °C
Storage Temperature: Ambient
MDL Number: MFCD02683110
CAS Number: 3133-81-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR