2-Fluoro-4-pyridone ≥97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 2-Fluoropyridin-4-ol, 2-Fluoro-4-hydroxypyridine

H31697.MD H31697.03
ALFAH31697.MDEA 0 SGD
ALFAH31697.MD ALFAH31697.03
2-Fluoro-4-pyridone ≥97%
2-Fluoro-4-pyridone

Purity: 97% CAS No.: 22282-69-5 Molecular Formula: C5H4FNO Formula Weight: 113.09 Melting Point: 146-149° MDL No.: MFCD04114159

Formula: C₅H₄FNO
MW: 113.09 g/mol
Melting Pt: 146…149 °C
Storage Temperature: Ambient
MDL Number: MFCD04114159
CAS Number: 22282-69-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR