2-Fluoro-6-pyridone ≥97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 2-Fluoro-6-hydroxypyridine, 6-Fluoropyridin-2-ol

H31686.MD H31686.03
ALFAH31686.MDEA 0 SGD
ALFAH31686.MD ALFAH31686.03
2-Fluoro-6-pyridone ≥97%
2-Fluoro-6-pyridone

Purity: 97% CAS No.: 50543-23-2 Molecular Formula: C5H4FNO Formula Weight: 113.09 Melting Point: 125-129° MDL No.: MFCD03092938

Formula: C₅H₄FNO
MW: 113.09 g/mol
Melting Pt: 125…129 °C
Storage Temperature: Ambient
MDL Number: MFCD03092938
CAS Number: 50543-23-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR