Niobium carbide ≥99% (metals basis)
Supplier: Thermo Scientific
|
Warning
|
12147.14
12147.22
ALFA12147.14EA
94.4
SGD
ALFA12147.14
ALFA12147.22
Niobium carbide ≥99% (metals basis)
Niobium carbide
Purity: 99+%
CAS No.: 12069-94-2
Molecular Formula: NbC
Formula Weight: 104.91
Physical Form:
Formula:
CNb MW: 104,91 g/mol Melting Pt: 3500 °C Density: 7,60 Storage Temperature: Ambient |
MDL Number:
MFCD00016237 CAS Number: 12069-94-2 EINECS: 235-117-8 UN: 3178 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...