Cyclohexanone cyanohydrin ≥98%
Supplier: Thermo Scientific
|
Danger
|
H26699.03
H26699.09
H26699.18
ALFAH26699.03EA
0
SGD
ALFAH26699.03
ALFAH26699.09
ALFAH26699.18
Cyclohexanone cyanohydrin ≥98%
Cyclohexanone cyanohydrin
Purity: 98%
CAS No.: 931-97-5
Molecular Formula: C7H11NO
Formula Weight: 125.17
Storage Temperature: 2°C to 8°C
Flash Point: 118°(244°F)
Refractive Index: 1.4690
Melting Point: 34-36°
Boiling Point: 132°/19mm
UN No.: 3439
MDL No.: MFCD00003818
Formula:
C₇H₁₁NO MW: 125,17 g/mol Boiling Pt: 132 °C (19 mmHg) Melting Pt: 34…36 °C Density: 1,031 Flash Pt: 118 °C (244 °F) |
MDL Number:
MFCD00003818 CAS Number: 931-97-5 EINECS: 213-246-0 UN: 3439 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...