(Pentamethylcyclopentadienyl)hafnium trichloride ≥98%

Supplier: Thermo Scientific
Danger

Synonyms: Pentamethylcyclopentadienylhafnium(IV) trichloride

H27010.03 H27010.06
ALFAH27010.03EA 194 SGD
ALFAH27010.03 ALFAH27010.06
(Pentamethylcyclopentadienyl)hafnium trichloride ≥98%
(Pentamethylcyclopentadienyl)hafnium trichloride

Purity: 98+% CAS No.: 75181-08-7 Molecular Formula: C10H15Cl3Hf Formula Weight: 420.08 Melting Point: 170-172° UN No.: 3261 MDL No.: MFCD00070461

Formula: C₁₀H₁₅Cl₃Hf
MW: 420,07 g/mol
Melting Pt: 170…172 °C
Storage Temperature: Ambient
MDL Number: MFCD00070461
CAS Number: 75181-08-7
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR