Bis(pentamethylcyclopentadienyl)hafnium(IV) dichloride ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Bis(pentamethylcyclopentadienyl)hafnium dichloride
H26912.03
H26912.06
ALFAH26912.03EA
0
SGD
ALFAH26912.03
ALFAH26912.06
Bis(pentamethylcyclopentadienyl)hafnium(IV) dichloride ≥98%
Bis(pentamethylcyclopentadienyl)hafnium(IV) dichloride
Purity: 98+%
CAS No.: 85959-83-7
Molecular Formula: C20H30Cl2Hf
Formula Weight: 519.86
Melting Point: 300°
MDL No.: MFCD00058887
Formula:
C₂₀H₃₀Cl₂Hf MW: 519,86 g/mol Melting Pt: 300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00058887 CAS Number: 85959-83-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...