3,5-Dichloro-2-pyridone ≥98%

Supplier: Thermo Scientific
Warning

L08309.03 L08309.06
ALFAL08309.03EA 165 SGD
ALFAL08309.03 ALFAL08309.06
3,5-Dichloro-2-pyridone ≥98%
3,5-Dichloro-2-pyridone

Purity: 98+% CAS No.: 5437-33-2 Molecular Formula: C5H3Cl2NO Formula Weight: 163.99 Melting Point: 178-182° Beilstein Reference: 1524971

Formula: C₅H₃Cl₂NO
MW: 163,99 g/mol
Melting Pt: 178…182 °C
Storage Temperature: Ambient
MDL Number: MFCD00015401
CAS Number: 5437-33-2
EINECS: 226-609-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR