(R)-(-)-1-(1-Naphthyl)ethyl isocyanate ≥97%

Supplier: Thermo Scientific
Danger

L12286.03 L12286.MD
ALFAL12286.03EA 284 SGD
ALFAL12286.03 ALFAL12286.MD
(R)-(-)-1-(1-Naphthyl)ethyl isocyanate ≥97%
(R)-(-)-1-(1-Naphthyl)ethyl isocyanate

Purity: 97% CAS No.: 42340-98-7 Molecular Formula: C13H11NO Formula Weight: 197.24 Storage Temperature: 2°C to 8°C Flash Point: 93°(199°F) Refractive Index: 1.6045 Boiling Point: 106-108°/0.16m Beilstein Reference: 4181805 Optical Rotation: -47° (c=3.5 in toluene) UN No.: 2206 MDL No.: MFCD00004015

Formula: C₁₃H₁₁NO
MW: 197,24 g/mol
Boiling Pt: 106…108 °C (0,16 m)
Density: 1,118
Flash Pt: 93 °C (199 °F)
MDL Number: MFCD00004015
CAS Number: 42340-98-7
EINECS: 255-759-2
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR