Ethyl-4-oxocyclohexanecarboxylate ≥97%

Supplier: Thermo Fisher Scientific
Warning

B20572.03 B20572.06
ALFAB20572.03EA 0 SGD
ALFAB20572.03 ALFAB20572.06
Ethyl-4-oxocyclohexanecarboxylate ≥97%
Ethyl 4-oxocyclohexanecarboxylate

Purity: 97% CAS No.: 17159-79-4 Molecular Formula: C9H14O3 Formula Weight: 170.21 Refractive Index: 1.4600 Boiling Point: 150-152°/40mm MDL No.: MFCD00013285

Formula: C₉H₁₄O₃
Boiling Pt: 150…152 °C (40 mmHg)
Density: 1,068
Storage Temperature: Ambient
MDL Number: MFCD00013285
CAS Number: 17159-79-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR