Borane tetrahydrofuran complex 1 mol/l in THF
Supplier: Thermo Scientific
|
Danger
|
Packaged under argon in resealable ChemSeal™ bottles.
43291.JQ
43291.JT
ALFA43291.JQEA
0
SGD
ALFA43291.JQ
ALFA43291.JT
Borane tetrahydrofuran complex 1 mol/l in THF
Borane tetrahydrofuran complex 1 mol/l in tetrahydrofuran
CAS No.: 14044-65-6
Molecular Formula: C4H11BO
Formula Weight: 85.94
Storage Temperature: 2°C to 8°C
Flash Point: -21°(-8°F)
Physical Form: Liquid
Density: 0.878
Sensitivity: Air and Moisture Sensitive
Appearance: Colourless
Beilstein Reference: 3668402
UN No.: 3148
MDL No.: MFCD00012429
Formula:
C₄H₁₁BO MW: 85,94 g/mol Density: 0,878 Flash Pt: –21 °C (–8 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00012429 CAS Number: 14044-65-6 EINECS: 237-881-8 UN: 3148 ADR: 4.3,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...