4-(4-Hydroxyphenyl)cyclohexanone ≥97%

Supplier: Thermo Scientific
Warning

H55767.03 H55767.06 H55767.14
ALFAH55767.03EA 52.5 SGD
ALFAH55767.03 ALFAH55767.06 ALFAH55767.14
4-(4-Hydroxyphenyl)cyclohexanone ≥97%
4-(4-Hydroxyphenyl)cyclohexanone

Purity: 97+% CAS No.: 105640-07-1 Molecular Formula: C12H14O2 Formula Weight: 190.24 Melting Point: 170-175° Beilstein Reference: 3269332

Formula: C₁₂H₁₄O₂
MW: 190,24 g/mol
Melting Pt: 170…175 °C
Storage Temperature: Ambient
MDL Number: MFCD00210693
CAS Number: 105640-07-1
EINECS: 423-920-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR