5-Bromo-2-methoxypyrimidine 95%

Supplier: Thermo Fisher Scientific
Danger

H31918.MD H31918.03
ALFAH31918.MDEA 0 SGD
ALFAH31918.MD ALFAH31918.03
5-Bromo-2-methoxypyrimidine 95%
5-Bromo-2-methoxypyrimidine

Purity: 95% CAS No.: 14001-66-2 Molecular Formula: C5H5BrN2O Formula Weight: 189.01 MDL No.: MFCD04973351

Formula: C₅H₅BrN₂O
MW: 189.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04973351
CAS Number: 14001-66-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR