4-Ethylphenylacetone ≥97%

Supplier: Thermo Scientific

A18810.06 A18810.03
ALFAA18810.06EA 0 SGD
ALFAA18810.06 ALFAA18810.03
4-Ethylphenylacetone ≥97%
4-Ethylphenylacetone

Purity: 97+% CAS No.: 75251-24-0 Molecular Formula: C11H14O Formula Weight: 162.23 Refractive Index: 1.5120

Formula: C₁₁H₁₄O
MW: 162,23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00210429
CAS Number: 75251-24-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR