1-(2-Thienyl)-1-propanone ≥98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
2-Propionylthiophene
L02318.06
L02318.14
L02318.22
ALFAL02318.06EA
63.4
SGD
ALFAL02318.06
ALFAL02318.14
ALFAL02318.22
1-(2-Thienyl)-1-propanone ≥98%
1-(2-Thienyl)-1-propanone
Purity: 98%
CAS No.: 13679-75-9
Molecular Formula: C7H8OS
Formula Weight: 140.20
Flash Point: 100°(212°F)
Refractive Index: 1.5530
Boiling Point: 228°
Beilstein Reference: 108934
MDL No.: MFCD00005446
Formula:
C₇H₈OS MW: 140,20 g/mol Boiling Pt: 228 °C Density: 1,124 Flash Pt: 100 °C (212 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005446 CAS Number: 13679-75-9 EINECS: 237-182-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...