4-Chloro-2-(trifluoromethyl)quinoline ≥98%
Supplier: Thermo Scientific
|
Warning
|
A10834.03
A10834.06
ALFAA10834.03EA
0
SGD
ALFAA10834.03
ALFAA10834.06
4-Chloro-2-(trifluoromethyl)quinoline ≥98%
4-Chloro-2-(trifluoromethyl)quinoline
Purity: 98%
CAS No.: 1701-24-2
Molecular Formula: C10H5ClF3N
Formula Weight: 231.61
Flash Point: 107°(225°F)
Melting Point: 31-33°
MDL No.: MFCD00153105
Formula:
C₁₀H₅ClF₃N MW: 231,61 g/mol Melting Pt: 31…33 °C Flash Pt: 107 °C (225 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00153105 CAS Number: 1701-24-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...