(12-Bromododecyl)phosphonic acid 95%

Supplier: Thermo Scientific
Warning

H33714.MD H33714.03
ALFAH33714.MDEA 314 SGD
ALFAH33714.MD ALFAH33714.03
(12-Bromododecyl)phosphonic acid 95%
(12-Bromododecyl)phosphonic acid
Formula: C₁₂H₂₆BrO₃P
MW: 329,21 g/mol
Melting Pt: 92…94 °C
Storage Temperature: Ambient
MDL Number: MFCD11046103
CAS Number: 202920-07-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR