Diethyl (hydroxymethyl)phosphonate tech. 85%, Technical Grade

Supplier: Thermo Scientific
Warning

B21215.06 B21215.14 B21215.22
ALFAB21215.06EA 0 SGD
ALFAB21215.06 ALFAB21215.14 ALFAB21215.22
Diethyl (hydroxymethyl)phosphonate tech. 85%, Technical Grade
Diethyl (hydroxymethyl)phosphonate

Purity: 85% CAS No.: 3084-40-0 Molecular Formula: C5H13O4P Formula Weight: 168.13 Flash Point: 110°(230°F) Refractive Index: 1.444 Boiling Point: 124-126°/3mm MDL No.: MFCD00014418

Formula: C₅H₁₃O₄P
MW: 168,13 g/mol
Boiling Pt: 124…126 °C (3 mmHg)
Density: 1,14
Flash Pt: 110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014418
CAS Number: 3084-40-0
EINECS: 221-391-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR