4-Hydroxybenzylideneacetone ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: 4-Hydroxybenzilideneacetone, 4-(4-Hydroxyphenyl)-3-buten-2-one, 4-(4-Hydroxyphenyl)but-3-en-2-one, 4-Hydroxybenzalacetone

A17605.14 A17605.22
ALFAA17605.14EA 0 SGD
ALFAA17605.14 ALFAA17605.22
4-Hydroxybenzylideneacetone ≥97%
4-Hydroxybenzylideneacetone

Purity: 97% CAS No.: 3160-35-8 Molecular Formula: C10H10O2 Formula Weight: 162.19 Melting Point: 106-111° Beilstein Reference: 2412544

Formula: C₁₀H₁₀O₂
MW: 162,19 g/mol
Melting Pt: 106…111 °C
Storage Temperature: Ambient
MDL Number: MFCD00016490
CAS Number: 3160-35-8
EINECS: 221-607-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR