Methyl-2-amino-5-chloro-3-iodobenzoate ≥98%

Supplier: Thermo Scientific
Warning

L20188.06 L20188.14
ALFAL20188.06EA 0 SGD
ALFAL20188.06 ALFAL20188.14
Methyl-2-amino-5-chloro-3-iodobenzoate ≥98%
Methyl 2-amino-5-chloro-3-iodobenzoate

Purity: 98+% CAS No.: 289039-84-5 Molecular Formula: C8H7ClINO2 Formula Weight: 311.51 Melting Point: 67-69° MDL No.: MFCD00673017

Formula: C₈H₇ClINO₂
MW: 311,51 g/mol
Melting Pt: 67…69 °C
Storage Temperature: Ambient
MDL Number: MFCD00673017
CAS Number: 289039-84-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR