4-Chloro-2-methyl-7-(trifluoromethyl)quinoline ≥97%

Supplier: Thermo Scientific
Warning

H50555.03 H50555.MD
ALFAH50555.03EA 551 SGD
ALFAH50555.03 ALFAH50555.MD
4-Chloro-2-methyl-7-(trifluoromethyl)quinoline ≥97%
4-Chloro-2-methyl-7-(trifluoromethyl)quinoline

Purity: 97% CAS No.: 18529-09-4 Molecular Formula: C11H7ClF3N Formula Weight: 245.63 MDL No.: MFCD00272334

Formula: C₁₁H₇ClF₃N
MW: 245,63 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00272334
CAS Number: 18529-09-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR