(±)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: rac-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl

H27585.MD H27585.03 H27585.06
ALFAH27585.MDEA 0 SGD
ALFAH27585.MD ALFAH27585.03 ALFAH27585.06
(±)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl ≥98%
(±)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl

Purity: 98% CAS No.: 153305-67-0 Molecular Formula: C48H40P2 Formula Weight: 678.79 Melting Point: 267-271° MDL No.: MFCD01311709

Formula: C₄₈H₄₀P₂
MW: 678,79 g/mol
Melting Pt: 267…271 °C
Storage Temperature: Ambient
MDL Number: MFCD01311709
CAS Number: 153305-67-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR