Benzylmagnesium chloride 1 M in MeTHF (2-methyltetrahydrofuran)

Supplier: Thermo Scientific
Danger

H51154.AP H51154.AE
ALFAH51154.APEA 2000 SGD
ALFAH51154.AP ALFAH51154.AE
Benzylmagnesium chloride 1 M in MeTHF (2-methyltetrahydrofuran)
Benzylmagnesium chloride

CAS No.: 6921-34-2 Molecular Formula: C7H7ClMg Formula Weight: 150.89 UN No.: 2924 MDL No.: MFCD00000469

Formula: C₇H₇ClMg
MW: 150,89 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00000469
CAS Number: 6921-34-2
EINECS: 230-039-0
UN: 2924
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR