2-(1,3,4-Oxadiazol-2-yl)phenol ≥98%
Supplier: Thermo Scientific
|
Warning
|
H50429.03
H50429.MD
ALFAH50429.03EA
561
SGD
ALFAH50429.03
ALFAH50429.MD
2-(1,3,4-Oxadiazol-2-yl)phenol ≥98%
2-(1,3,4-Oxadiazol-2-yl)phenol
Purity: 98%
CAS No.: 1008-65-7
Molecular Formula: C8H6N2O2
Formula Weight: 162.15
MDL No.: MFCD00020713
Formula:
C₈H₆N₂O₂ MW: 162,15 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00020713 CAS Number: 1008-65-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...