4'-Bromo-3'-(trifluoromethyl)acetanilide 97

Supplier: Thermo Fisher Scientific
Warning

Synonyms: N-(4-Bromo-3-(trifluoromethyl)phenyl)acetamide

A19939.09 A19939.88
ALFAA19939.09EA 0 SGD
ALFAA19939.09 ALFAA19939.88
4'-Bromo-3'-(trifluoromethyl)acetanilide 97
4'-Bromo-3'-(trifluoromethyl)acetanilide

Purity: 97% CAS No.: 41513-05-7 Molecular Formula: C9H7BrF3NO Formula Weight: 282.07 Melting Point: 116-119° MDL No.: MFCD00052361

Formula: C₉H₇BrF₃NO
MW: 282.07 g/mol
Melting Pt: 116…119 °C
Storage Temperature: Ambient
MDL Number: MFCD00052361
CAS Number: 41513-05-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR