1-(2-Fluorophenyl)pyrrole ≥98%
Supplier: Thermo Scientific
H33247.03
H33247.06
ALFAH33247.03EA
0
SGD
ALFAH33247.03
ALFAH33247.06
1-(2-Fluorophenyl)pyrrole ≥98%
1-(2-Fluorophenyl)pyrrole
Purity: 98%
CAS No.: 89096-77-5
Molecular Formula: C10H8FN
Formula Weight: 161.18
Refractive Index: 1.5105
Boiling Point: 64°/1.1mm
MDL No.: MFCD02665231
Formula:
C₁₀H₈FN MW: 161,18 g/mol Boiling Pt: 64 °C (1,1 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD02665231 CAS Number: 89096-77-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...