2-Fluoro-DL-phenylalanine ≥98%

Supplier: Thermo Scientific

Synonyms: 2-Amino-3-(2-fluorophenyl)propanoic acid

L07654.03 L07654.06
ALFAL07654.03EA 77.1 SGD
ALFAL07654.03 ALFAL07654.06
2-Fluoro-DL-phenylalanine ≥98%
2-Fluoro-DL-phenylalanine

Purity: 98% CAS No.: 2629-55-2 Molecular Formula: C9H10FNO2 Formula Weight: 183.18 Melting Point: ca 245° dec. Beilstein Reference: 3201186

Formula: C₉H₁₀FNO₂
MW: 183,18 g/mol
Melting Pt: ∼ 245 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00004271
CAS Number: 2629-55-2
EINECS: 220-105-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR