4'-(Diethylamino)acetophenone 99%
Supplier: Thermo Scientific
Synonyms:
1-(4-(Diethylamino)phenyl)ethanone
L13331.03
L13331.06
ALFAL13331.03EA
0
SGD
ALFAL13331.03
ALFAL13331.06
4'-(Diethylamino)acetophenone 99%
4'-(Diethylamino)acetophenone
Purity: 99%
CAS No.: 5520-66-1
Molecular Formula: C12H17NO
Formula Weight: 191.27
Melting Point: 47-49°
Beilstein Reference: 2093988
Formula:
C₁₂H₁₇NO MW: 191,27 g/mol Melting Pt: 47…49 °C Storage Temperature: Ambient |
MDL Number:
MFCD00043651 CAS Number: 5520-66-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...