Methyl benzo[b]thiophene-2-carboxylate ≥98%
Supplier: Thermo Scientific
A13769.03
A13769.06
ALFAA13769.03EA
0
SGD
ALFAA13769.03
ALFAA13769.06
Methyl benzo[b]thiophene-2-carboxylate ≥98%
Methyl benzo[b]thiophene-2-carboxylate
Purity: 98%
CAS No.: 22913-24-2
Molecular Formula: C10H8O2S
Formula Weight: 192.24
Melting Point: 69-70°
Beilstein Reference: 142166
Formula:
C₁₀H₈O₂S MW: 192,24 g/mol Melting Pt: 69…70 °C Storage Temperature: Ambient |
MDL Number:
MFCD00067791 CAS Number: 22913-24-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...