(R)-(+)-2-Methyl-CBS-oxazaborolidine monohydrate 94%
Supplier: Thermo Scientific
Synonyms:
(R)-2-Methyl-CBS-oxazaborolidine monohydrate, (R)-1-Methyl-3,3-diphenylhexahydropyrrolo[1,2-c][1,3,2]oxazaborole, (R)-3,3-Diphenyl-1-methylpyrrolidino[1,2-c]-1,3,2-oxazaborole, α,α-Diphenyl-D-prolinolmethylboronic acid cyclamide ester
Purity: 94%
CAS No.: 112022-83-0
Molecular Formula: C18H20BNO.H2O
Formula Weight: 295.19 (277.17anhy)
Beilstein Reference: 9059874
Optical Rotation: +74° (c=1 in methanol)
Formula:
C₁₈H₂₀BNO·1H₂O MW: 295,19 g/mol (277,17 anhy) Storage Temperature: Ambient |
MDL Number:
MFCD00078440 CAS Number: 112022-83-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...