Membrane filters, PORAFIL® TE
Supplier: MACHEREY-NAGEL
670020013 670020025 670020047 670020050 670020090 670020100 670020142 670020220 670045013 670045025 670045047 670045050 670045090 670045100 670045142 670045220 670100013 670100025 670100047 670100050 670100090 670100100 670100142 670100220 670300013 670300025 670300047 670300050 670300090 670300100 670300142 670300220
MANA670020013EA 0 SGD
MANA670020013 MANA670020025 MANA670020047 MANA670020050 MANA670020090 MANA670020100 MANA670020142 MANA670020220 MANA670045013 MANA670045025 MANA670045047 MANA670045050 MANA670045090 MANA670045100 MANA670045142 MANA670045220 MANA670100013 MANA670100025 MANA670100047 MANA670100050 MANA670100090 MANA670100100 MANA670100142 MANA670100220 MANA670300013 MANA670300025 MANA670300047 MANA670300050 MANA670300090 MANA670300100 MANA670300142 MANA670300220
Membrane filters, PORAFIL® TE
Filters Membrane Filters
Membranes of PTFE are particularly suited for aggressive media, they are chemically inert towards aqueous and organic solutions, concentrated acids and bases.
  • Hydrophobic
  • Can be used at temperatures up to 145 °C