2,6-Dimethoxy-3-pyridineboronic acid 95%
Supplier: Thermo Scientific
|
Warning
|
L20389.03
L20389.06
L20389.MD
ALFAL20389.03EA
321
SGD
ALFAL20389.03
ALFAL20389.06
ALFAL20389.MD
2,6-Dimethoxy-3-pyridineboronic acid 95%
2,6-Dimethoxy-3-pyridineboronic acid
Purity: 95%
CAS No.: 221006-70-8
Molecular Formula: C7H10BNO4
Formula Weight: 182.97
Melting Point: 128-130°
Beilstein Reference: 9391039
Formula:
C₇H₁₀BNO₄ MW: 182,97 g/mol Melting Pt: 128…130 °C Storage Temperature: Ambient |
MDL Number:
MFCD03788239 CAS Number: 221006-70-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...